The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001112699.1 | NP_001106169.1 | 9 - 10 | 941 | 82 |
GenBank mRNA | AB013464.1 | - | 9 - 10 | 862 | 82 |
AK078502.1 | - | - | 851 | 82 | |
AY126516.1 | - | 9 - 10 | 910 | 82 | |
BC057974.1 | - | 9 - 10 | 825 | 82 |