Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ces2f
Entrez Gene ID
71903
Gene Name
carboxylesterase 2F
Gene Aliases
2310038E17Rik, A430088E12
Chromosome Location
Chr.8: 104947355 - 104961130 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079865.2 NP_001073334.2 10 - 11 1455 108
GenBank mRNA AK040349.1 - 10 - 11 1457 108
BC117742.1 - 10 - 11 1455 108
BC117743.1 - 8 - 9 1062 108

View Full Product Details