The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013602.3 | NP_038630.1 | 1 - 2 | 251 | 62 |
GenBank mRNA | AK007983.1 | - | 1 - 2 | 100 | 62 |
AK011040.1 | - | 1 - 2 | 100 | 62 | |
AK018727.1 | - | 1 - 2 | 114 | 62 | |
BC027262.1 | - | 1 - 2 | 129 | 62 | |
BC036990.1 | - | - | 96 | 62 | |
BC109369.1 | - | - | 610 | 62 |