Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rgs9
Entrez Gene ID
19739
Gene Name
regulator of G-protein signaling 9
Gene Aliases
RGS9-1, Rgs9-2
Chromosome Location
Chr.11: 109225348 - 109298181 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001165934.1 NP_001159406.1 15 - 16 1376 65
NM_011268.2 NP_035398.2 15 - 16 1376 65
GenBank mRNA AF011358.1 - 15 - 16 1261 65
AF125046.1 - 15 - 16 1332 65
AK138159.1 - 15 - 16 1286 65
BC129898.1 - 7 - 8 654 65
BC129899.1 - 15 - 16 1242 65

View Full Product Details