Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nuak1
Entrez Gene ID
77976
Gene Name
NUAK family, SNF1-like kinase, 1
Gene Aliases
AU014801, AW494241, B230104P22Rik, Omphk1
Chromosome Location
Chr.10: 84370900 - 84440471 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004363.1 NP_001004363.1 5 - 6 821 85
GenBank mRNA AB182364.1 - 4 - 5 458 85
BC040467.1 - 1 - 2 117 85
BC082328.1 - 5 - 6 821 85

View Full Product Details