Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Psmd12
Entrez Gene ID
66997
Gene Name
proteasome (prosome, macropain) 26S subunit, non-ATPase, 12
Gene Aliases
1500002F15Rik, AI480719, P55
Chromosome Location
Chr.11: 107479528 - 107498036 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025894.2 NP_080170.1 4 - 5 446 78
GenBank mRNA AK005114.1 - 4 - 5 481 78
AK007619.1 - 4 - 5 491 78
AK007680.1 - 4 - 5 420 78
AK008646.1 - 4 - 5 447 78
AK009969.1 - 4 - 5 449 78
AK011806.1 - 4 - 5 470 78
AK017843.1 - 4 - 5 463 78
AK150910.1 - 4 - 5 470 78
AK151242.1 - 4 - 5 447 78
AK152041.1 - 4 - 5 470 78
AK152240.1 - 4 - 5 478 78
AK160283.1 - 4 - 5 468 78
AK162779.1 - - 447 78
AK167236.1 - 4 - 5 454 78
BC004694.1 - 4 - 5 464 78

View Full Product Details