Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cyp24a1
Entrez Gene ID
13081
Gene Name
cytochrome P450, family 24, subfamily a, polypeptide 1
Gene Aliases
24-OHase, CP24, Cyp24
Chromosome Location
Chr.2: 170482965 - 170497138 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009996.3 NP_034126.1 5 - 6 1094 103
GenBank mRNA AK143984.1 - 5 - 6 1102 103
AK159109.1 - 5 - 6 1102 103
AK159481.1 - 5 - 6 1102 103
AK159527.1 - 5 - 6 1102 103
AK159852.1 - 5 - 6 1102 103
AK159954.1 - 5 - 6 1102 103
AK159976.1 - 5 - 6 1102 103
AK164697.1 - 5 - 6 1102 103
BC109139.1 - 5 - 6 1086 103
BC109140.1 - 5 - 6 1086 103
D49438.1 - 5 - 6 1032 103
D89669.1 - 5 - 6 1094 103

View Full Product Details