Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zdbf2
Entrez Gene ID
73884
Gene Name
zinc finger, DBF-type containing 2
Gene Aliases
4930431J08Rik, 9330107J05Rik
Chromosome Location
Chr.1: 63200358 - 63314576 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001267872.1 NP_001254801.1 4 - 5 505 149
GenBank mRNA AK132808.1 - 4 - 5 506 149

View Full Product Details