Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Aox2
Entrez Gene ID
213043
Gene Name
aldehyde oxidase 2
Gene Aliases
Aoh3, Aox3l1
Chromosome Location
Chr.1: 58276141 - 58379264 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008419.2 NP_001008419.1 21 - 22 2458 98
GenBank mRNA AY187018.2 - 21 - 22 2458 98
AY665589.1 - 21 - 22 2365 98
BC150826.1 - 21 - 22 2385 98

View Full Product Details