Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ppil3
Entrez Gene ID
70225
Gene Name
peptidylprolyl isomerase (cyclophilin)-like 3
Gene Aliases
2310076N22Rik, 2510026K04Rik, Cyp10l
Chromosome Location
Chr.1: 58430993 - 58445527 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001285826.1 NP_001272755.1 6 - 7 1085 85
NM_001285827.1 NP_001272756.1 6 - 7 750 85
NM_027351.3 NP_081627.1 6 - 7 564 85
GenBank mRNA AF315803.1 - 6 - 7 698 85
AK010207.1 - 6 - 7 568 85
BB687332.1 - 1 - 2 56 85
BC060993.1 - 6 - 7 532 85
BC061645.1 - 6 - 7 1028 85

View Full Product Details