The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001161798.1 | NP_001155270.1 | 7 - 8 | 1304 | 61 |
NM_010840.3 | NP_034970.2 | 7 - 8 | 1227 | 61 | |
NR_027809.1 | 7 - 8 | 1304 | 61 | ||
GenBank mRNA | AK030192.1 | - | 7 - 8 | 1222 | 61 |
AK149795.1 | - | 7 - 8 | 1289 | 61 | |
AK150159.1 | - | 7 - 8 | 1305 | 61 | |
BC051017.1 | - | 7 - 8 | 1206 | 61 | |
BC052466.1 | - | 7 - 8 | 1271 | 61 |