Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rasef
Entrez Gene ID
242505
Gene Name
RAS and EF hand domain containing
Gene Aliases
BC030882, RAB45
Chromosome Location
Chr.4: 73714577 - 73791401 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017427.1 NP_001017427.1 17 - 18 1988 60
GenBank mRNA AK132691.1 - 17 - 18 1968 60
BC030882.1 - 13 - 14 1381 60
BC040766.1 - 13 - 14 1370 60
BC125539.1 - 16 - 17 1802 60
BC137774.1 - 16 - 17 1802 60
BC144901.1 - 16 - 17 1738 60

View Full Product Details