Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Ccbe1
- Entrez Gene ID
-
320924
- Gene Name
- collagen and calcium binding EGF domains 1
- Gene Aliases
-
4933426F18Rik, 9430093N24Rik, mKIAA1983
- Chromosome Location
-
Chr.18: 66056855 - 66291838 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK016929.1 |
- |
- |
515 |
88 |
View Full Product Details