Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gramd3
Entrez Gene ID
107022
Gene Name
GRAM domain containing 3
Gene Aliases
9030613F08Rik, 9130427A09Rik, AW061306, NS3TP2
Chromosome Location
Chr.18: 56387485 - 56503792 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026240.2 NP_080516.2 2 - 3 351 72
GenBank mRNA AK018551.1 - 3 - 4 601 72
AK018691.1 - 2 - 3 337 72
AK038289.1 - 1 - 2 392 72
AK161329.1 - 2 - 3 284 72
AK164050.1 - 2 - 3 336 72
BC057978.1 - 2 - 3 326 72

View Full Product Details