Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Phax
Entrez Gene ID
56698
Gene Name
phosphorylated adaptor for RNA export
Gene Aliases
2810055C14Rik, 4933427L19Rik, AU018701, AU018854, D18Ertd65e, Rnuxa, p55
Chromosome Location
Chr.18: 56562569 - 56587712 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001162989.1 NP_001156461.1 2 - 3 744 64
NM_019996.4 NP_064380.3 2 - 3 854 64
GenBank mRNA AJ276504.1 - 2 - 3 685 64
AK016958.1 - 2 - 3 746 64
BC016530.1 - 2 - 3 701 64
BC049590.1 - 2 - 3 700 64
BC061110.1 - 2 - 3 714 64

View Full Product Details