The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001038608.2 | NP_001033697.2 | 1 - 2 | 108 | 157 |
NM_025299.3 | NP_079575.1 | 1 - 2 | 108 | 157 | |
GenBank mRNA | BY065764.1 | - | 1 - 2 | 109 | 157 |