The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001252481.1 | NP_001239410.1 | 10 - 11 | 1724 | 94 |
NM_010754.5 | NP_034884.2 | 10 - 11 | 1613 | 94 | |
GenBank mRNA | AF005743.1 | - | 9 - 10 | 1341 | 94 |
AK007817.1 | - | 10 - 11 | 1390 | 94 | |
AK088249.1 | - | 1 - 2 | 73 | 94 | |
AY334552.1 | - | 8 - 9 | 1192 | 94 | |
BC021342.1 | - | 10 - 11 | 1412 | 94 | |
BC089184.1 | - | 10 - 11 | 1724 | 94 | |
U60530.1 | - | 9 - 10 | 1341 | 94 |