Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hsd17b4
Entrez Gene ID
15488
Gene Name
hydroxysteroid (17-beta) dehydrogenase 4
Gene Aliases
17-beta-HSD, 17[b]-HSD, 4, DBP, MFE-2, MFP2, MPF-2, Mfp-2, perMFE-2
Chromosome Location
Chr.18: 50128201 - 50196270 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008292.4 NP_032318.2 6 - 7 499 74
GenBank mRNA AK004866.1 - 6 - 7 499 74
AK088381.1 - 6 - 7 474 74
AK147182.1 - 6 - 7 471 74
AK152532.1 - 6 - 7 394 74
AK161655.1 - 6 - 7 394 74
AK166801.1 - 6 - 7 412 74
AK167060.1 - 6 - 7 449 74
AK169077.1 - 6 - 7 401 74
BC022175.1 - 6 - 7 372 74
X89998.1 - 6 - 7 360 74

View Full Product Details