Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dctn4
Entrez Gene ID
67665
Gene Name
dynactin 4
Gene Aliases
1110001K06Rik, 4930547K17Rik, C130039E17Rik, p62
Chromosome Location
Chr.18: 60526159 - 60558762 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026302.3 NP_080578.1 3 - 4 397 95
GenBank mRNA AK016059.1 - 3 - 4 451 95
AK034292.1 - 3 - 4 434 95
AK050896.1 - 3 - 4 401 95
AK141855.1 - 3 - 4 398 95
AK160813.1 - 3 - 4 403 95
AK163240.1 - 3 - 4 419 95
AK168484.1 - 3 - 4 403 95
BC006677.1 - 3 - 4 360 95
BC034725.1 - 3 - 4 397 95

View Full Product Details