Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tmod4
Entrez Gene ID
50874
Gene Name
tropomodulin 4
Gene Aliases
MTMOD, Sk-Tmod
Chromosome Location
Chr.3: 95124117 - 95129208 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016712.3 NP_057921.1 6 - 7 723 66
GenBank mRNA AF177174.1 - 5 - 6 642 66
AK144408.1 - 6 - 7 724 66
AK144535.1 - 6 - 7 724 66
BC068020.1 - 5 - 6 659 66

View Full Product Details