Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Ssbp3
- Entrez Gene ID
-
72475
- Gene Name
- single-stranded DNA binding protein 3
- Gene Aliases
-
2610021L12Rik, 2610200M23Rik, 5730488C10Rik, AI854733, AW551939, LAST, SSDP1
- Chromosome Location
-
Chr.4: 106911470 - 107049694 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK011509.1 |
- |
1 - 2 |
65 |
80 |
View Full Product Details