Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Atg9a
Entrez Gene ID
245860
Gene Name
autophagy related 9A
Gene Aliases
AU019532, Apg9l1, Atg9, Atg9l1
Chromosome Location
Chr.1: 75180861 - 75192010 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003917.4 NP_001003917.2 5 - 6 488 90
NM_001288612.1 NP_001275541.1 5 - 6 491 90
NM_001288613.1 NP_001275542.1 5 - 6 488 90
NR_109938.1 5 - 6 491 90
GenBank mRNA AK043624.1 - 5 - 6 542 90
AM085510.1 - 5 - 6 401 90
BC056488.1 - 5 - 6 401 90
BC079884.1 - 5 - 6 436 90
BC082307.1 - 3 - 4 194 90

View Full Product Details