The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001003917.4 | NP_001003917.2 | 9 - 10 | 1693 | 65 |
NM_001288612.1 | NP_001275541.1 | 9 - 10 | 1696 | 65 | |
NM_001288613.1 | NP_001275542.1 | 9 - 10 | 1941 | 65 | |
GenBank mRNA | AM085510.1 | - | 9 - 10 | 1606 | 65 |
BC034139.1 | - | 2 - 3 | 382 | 65 | |
BC056488.1 | - | 9 - 10 | 1854 | 65 | |
BC082307.1 | - | 7 - 8 | 1399 | 65 |