Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ankrd35
Entrez Gene ID
213121
Gene Name
ankyrin repeat domain 35
Gene Aliases
4732436F15Rik
Chromosome Location
Chr.3: 96670131 - 96691034 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081139.1 NP_001074608.1 11 - 12 3147 155
GenBank mRNA AK028691.1 - 11 - 12 2937 155
AK046624.1 - 11 - 12 2957 155
BC138276.1 - 11 - 12 2952 155
BC145228.1 - 11 - 12 2956 155
BC145750.1 - 11 - 12 2952 155
BC157973.1 - 11 - 12 2952 155

View Full Product Details