Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ankrd35
Entrez Gene ID
213121
Gene Name
ankyrin repeat domain 35
Gene Aliases
4732436F15Rik
Chromosome Location
Chr.3: 96670131 - 96691034 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081139.1 NP_001074608.1 1 - 2 315 61
GenBank mRNA AK039772.1 - 1 - 2 70 61
AK046624.1 - 1 - 2 120 61
BC138276.1 - 1 - 2 117 61
BC145228.1 - 1 - 2 117 61
BC145750.1 - 1 - 2 117 61
BC157973.1 - 1 - 2 117 61

View Full Product Details