Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ankrd35
Entrez Gene ID
213121
Gene Name
ankyrin repeat domain 35
Gene Aliases
4732436F15Rik
Chromosome Location
Chr.3: 96670131 - 96691034 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081139.1 NP_001074608.1 5 - 6 666 84
GenBank mRNA AK028691.1 - 5 - 6 456 84
AK039772.1 - 5 - 6 421 84
AK046624.1 - 5 - 6 475 84
BC138276.1 - 5 - 6 468 84
BC145228.1 - 5 - 6 472 84
BC145750.1 - 5 - 6 468 84
BC157973.1 - 5 - 6 468 84

View Full Product Details