Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Capn11
Entrez Gene ID
268958
Gene Name
calpain 11
Gene Aliases
-
Chromosome Location
Chr.17: 45630191 - 45660090 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013767.2 NP_001013789.1 6 - 7 809 77
GenBank mRNA AY578330.1 - 6 - 7 809 77

View Full Product Details