Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fnbp4
Entrez Gene ID
55935
Gene Name
formin binding protein 4
Gene Aliases
FBP30, Fnbp30, mKIAA1014
Chromosome Location
Chr.2: 90745370 - 90781021 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018828.2 NP_061298.1 16 - 17 3231 78
GenBank mRNA AK013152.1 - 2 - 3 294 78
AK084018.1 - 16 - 17 3023 78
AK129263.1 - 15 - 16 4102 78
U40750.2 - 16 - 17 3231 78

View Full Product Details