Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Trim6
Entrez Gene ID
94088
Gene Name
tripartite motif-containing 6
Gene Aliases
C430046K18Rik, D7Ertd684e
Chromosome Location
Chr.7: 104218783 - 104235152 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013616.2 NP_001013637.1 1 - 2 87 69
GenBank mRNA AK033396.1 - 1 - 2 67 69
AK040000.1 - 1 - 2 90 69
AK049238.1 - 1 - 2 67 69
BB634731.1 - 1 - 2 87 69

View Full Product Details