The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019439.3 | NP_062312.3 | 22 - 23 | 3026 | 70 |
GenBank mRNA | AF008649.1 | - | 22 - 23 | 2910 | 70 |
AF120255.1 | - | 17 - 18 | 2354 | 70 | |
AK038844.1 | - | - | 2066 | 70 | |
AK156397.1 | - | 20 - 21 | 2760 | 70 | |
BC054735.1 | - | 22 - 23 | 3026 | 70 | |
BC056990.1 | - | 17 - 18 | 2403 | 70 |