Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Stap2
Entrez Gene ID
106766
Gene Name
signal transducing adaptor family member 2
Gene Aliases
AW049765, STAP-2
Chromosome Location
Chr.17: 55997074 - 56005643 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145934.2 NP_666046.1 2 - 3 254 104
NR_102313.1 2 - 3 254 104
GenBank mRNA AK050131.1 - 2 - 3 234 104
AK050172.1 - 2 - 3 222 104
BC026642.1 - 2 - 3 228 104

View Full Product Details