Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lrrc32
Entrez Gene ID
434215
Gene Name
leucine rich repeat containing 32
Gene Aliases
AI426318, D11S833Eh, D7H11S833E, EG434215, Garp
Chromosome Location
Chr.7: 98489301 - 98501831 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113379.1 NP_001106850.1 1 - 2 84 62

View Full Product Details