Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- March7
- Entrez Gene ID
-
57438
- Gene Name
- membrane-associated ring finger (C3HC4) 7
- Gene Aliases
-
Axo, Axot, Gtrgeo17
- Chromosome Location
-
Chr.2: 60209892 - 60249211 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC025029.1 |
- |
1 - 2 |
111 |
133 |
View Full Product Details