The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_028142.1 | 1 - 2 | 288 | 65 | |
GenBank mRNA | BC019188.1 | - | 1 - 2 | 270 | 65 |
BC044737.1 | - | 1 - 2 | 252 | 65 | |
BC044781.2 | - | 1 - 2 | 252 | 65 | |
BC054825.1 | - | 1 - 2 | 247 | 65 |