Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tpcn2
Entrez Gene ID
233979
Gene Name
two pore segment channel 2
Gene Aliases
BC025890, D830047E22Rik
Chromosome Location
Chr.7: 145181633 - 145283987 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_146206.4 NP_666318.2 13 - 14 1205 74
GenBank mRNA AK040854.1 - 13 - 14 1233 74
AK052930.1 - 13 - 14 1206 74
AK089490.1 - 13 - 14 1200 74
AK090059.1 - 13 - 14 74
AK170863.1 - 13 - 14 1201 74
BC043472.1 - 9 - 10 783 74
BC141195.1 - 13 - 14 1289 74

View Full Product Details