Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ppapdc1a
Entrez Gene ID
381925
Gene Name
phosphatidic acid phosphatase type 2 domain containing 1A
Gene Aliases
C030048B12Rik, Gm1090
Chromosome Location
Chr.7: 129257094 - 129391307 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080963.1 NP_001074432.1 4 - 5 342 79
GenBank mRNA AK081292.1 - 4 - 5 407 79
BC120497.1 - 4 - 5 342 79

View Full Product Details