Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Harbi1
Entrez Gene ID
241547
Gene Name
harbinger transposase derived 1
Gene Aliases
D230010M03Rik
Chromosome Location
Chr.2: 91710855 - 91721567 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178724.4 NP_848839.2 1 - 2 299 107
GenBank mRNA AK045343.1 - 1 - 2 289 107
AK080671.1 - 1 - 2 328 107
AK084226.1 - 1 - 2 333 107
AK147045.1 - 1 - 2 339 107
BC094315.1 - 1 - 2 299 107

View Full Product Details