Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gm26705
Entrez Gene ID
381967
Gene Name
predicted gene, 26705
Gene Aliases
-
Chromosome Location
Chr.7: 99260481 - 99267218 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_103487.1 2 - 3 141 131
GenBank mRNA BC050000.1 - 2 - 3 141 131


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BQ929997.1 - 140 Chr. - - [-] on Build 131


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream monoacylglycerol O-acyltransferase 2 Mogat2 DGAT2L5, MGAT2, Mgat1l -25.0kb Sense Chr. - 99219084 - 99238611 on Build GRCm38
Downstream microtubule-associated protein 6 Map6 2810411E12Rik, Map-6, Mtap6, STOP 976.0 Anti-sense Chr. - 99267447 - 99337137 on Build GRCm38

View Full Product Details