The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080711.2 | NP_001074180.1 | 5 - 6 | 915 | 86 |
GenBank mRNA | DQ365828.1 | - | 5 - 6 | 915 | 86 |