Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zbtb10
Entrez Gene ID
229055
Gene Name
zinc finger and BTB domain containing 10
Gene Aliases
4832414A18, AU021107, RINZF
Chromosome Location
Chr.3: 9250567 - 9285333 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177660.3 NP_808328.2 2 - 3 2401 109
GenBank mRNA AK048280.1 - 1 - 2 862 109

View Full Product Details