The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001177950.1 | NP_001171421.1 | 1 - 2 | 242 | 151 |
NM_001177951.1 | NP_001171422.1 | 1 - 2 | 242 | 151 | |
NM_001177953.1 | NP_001171424.1 | 1 - 2 | 242 | 151 | |
NM_001177954.1 | NP_001171425.1 | 1 - 2 | 242 | 151 | |
NM_011285.2 | NP_035415.1 | 1 - 2 | 242 | 151 | |
GenBank mRNA | AF044677.1 | - | 1 - 2 | 160 | 151 |
AJ238396.1 | - | 1 - 2 | 154 | 151 | |
AK017192.1 | - | 1 - 2 | 174 | 151 | |
AK029864.1 | - | 1 - 2 | 368 | 151 | |
AK038652.1 | - | 1 - 2 | 206 | 151 | |
AK081631.1 | - | 1 - 2 | 181 | 151 | |
AK136298.1 | - | 1 - 2 | 173 | 151 |