Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hspa4
Entrez Gene ID
15525
Gene Name
heat shock protein 4
Gene Aliases
70kDa, AI317151, APG-2, Hsp110, Hsp70RY, mKIAA4025
Chromosome Location
Chr.11: 53259814 - 53300479 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008300.3 NP_032326.3 14 - 15 2122 65
GenBank mRNA AK053601.1 - 14 - 15 2056 65
AK054211.1 - 7 - 8 844 65
AK133772.1 - 14 - 15 2065 65
AK148883.1 - 3 - 4 397 65
AK155305.1 - 14 - 15 2039 65
AK166102.1 - 14 - 15 2105 65
AK220167.1 - 14 - 15 2073 65
BC003770.1 - 14 - 15 2028 65
BC003950.1 - 14 - 15 2017 65
CA316420.1 - 3 - 4 417 65

View Full Product Details