Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ankdd1b
Entrez Gene ID
271144
Gene Name
ankyrin repeat and death domain containing 1B
Gene Aliases
9330128J19Rik
Chromosome Location
Chr.13: 96415030 - 96471354 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042714.1 NP_001036179.1 9 - 10 1019 96
GenBank mRNA AK033386.1 - 3 - 4 259 96
AK033960.1 - 2 - 3 213 96

View Full Product Details