Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ankdd1b
Entrez Gene ID
271144
Gene Name
ankyrin repeat and death domain containing 1B
Gene Aliases
9330128J19Rik
Chromosome Location
Chr.13: 96415030 - 96471354 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042714.1 NP_001036179.1 11 - 12 1213 68
GenBank mRNA AK033386.1 - 5 - 6 453 68
AK033960.1 - 4 - 5 407 68
BC112417.1 - 11 - 12 1210 68

View Full Product Details