Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nuf2
Entrez Gene ID
66977
Gene Name
NUF2, NDC80 kinetochore complex component, homolog (S. cerevisiae)
Gene Aliases
2410003C07Rik, AU044112, C85691, Cdca1, NUF2R
Chromosome Location
Chr.1: 169497934 - 169531464 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_023284.3 NP_075773.2 9 - 10 821 129
GenBank mRNA AF326732.1 - 10 - 11 862 129
AK010351.1 - 9 - 10 743 129
AK012011.1 - 9 - 10 755 129
AK132467.1 - 9 - 10 822 129
AK162249.1 - 10 - 11 857 129
BC020026.1 - 10 - 11 830 129

View Full Product Details