Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nuf2
Entrez Gene ID
66977
Gene Name
NUF2, NDC80 kinetochore complex component, homolog (S. cerevisiae)
Gene Aliases
2410003C07Rik, AU044112, C85691, Cdca1, NUF2R
Chromosome Location
Chr.1: 169497934 - 169531464 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_023284.3 NP_075773.2 7 - 8 666 98
GenBank mRNA AF326732.1 - 8 - 9 707 98
AK010351.1 - 7 - 8 588 98
AK012011.1 - 7 - 8 600 98
AK089939.1 - 8 - 9 694 98
AK132467.1 - 7 - 8 667 98
AK162249.1 - 8 - 9 702 98
BC020026.1 - 8 - 9 675 98

View Full Product Details