Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nucks1
Entrez Gene ID
98415
Gene Name
nuclear casein kinase and cyclin-dependent kinase substrate 1
Gene Aliases
2700010L10Rik, 8430423A01Rik, AI647518, C78391, Nucks
Chromosome Location
Chr.1: 131910458 - 131936321 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145804.1 NP_001139276.1 3 - 4 448 135
NM_175294.3 NP_780503.2 3 - 4 448 135
GenBank mRNA AB049825.1 - 3 - 4 427 135
AK078816.1 - 3 - 4 372 135
BC039951.1 - 3 - 4 360 135

View Full Product Details