The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001024846.1 | NP_001020017.1 | 4 - 5 | 273 | 102 |
GenBank mRNA | AK028351.1 | - | 4 - 5 | 276 | 102 |
AK147890.1 | - | 5 - 6 | 769 | 102 | |
BC065692.1 | - | 1 - 2 | 377 | 102 |