The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011968.3 | NP_036098.1 | 2 - 3 | 266 | 74 |
GenBank mRNA | AF060087.1 | - | 2 - 3 | 175 | 74 |
AK088143.1 | - | 2 - 3 | 204 | 74 | |
AK161662.1 | - | 2 - 3 | 205 | 74 | |
BC086685.1 | - | 2 - 3 | 193 | 74 | |
BY340104.1 | - | 2 - 3 | 316 | 74 | |
U60288.1 | - | 2 - 3 | 208 | 74 |