Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tnnt2
Entrez Gene ID
21956
Gene Name
troponin T2, cardiac
Gene Aliases
Tnt, cTnT
Chromosome Location
Chr.1: 135836316 - 135852268 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130174.2 NP_001123646.1 6 - 7 311 73
NM_001130175.2 NP_001123647.4 6 - 7 307 73
NM_001130176.1 NP_001123648.1 4 - 5 198 73
NM_001130177.2 NP_001123649.1 5 - 6 299 73
NM_001130178.2 NP_001123650.1 5 - 6 281 73
NM_001130179.2 NP_001123651.4 5 - 6 277 73
NM_001130180.2 NP_001123652.1 4 - 5 269 73
NM_001130181.2 NP_001123653.4 4 - 5 265 73
NM_011619.3 NP_035749.1 4 - 5 269 73
GenBank mRNA AK146945.1 - 6 - 7 256 73
AK163786.1 - 5 - 6 244 73
AK168393.1 - 5 - 6 244 73
BC063753.1 - 6 - 7 287 73
BQ554639.1 - 5 - 6 217 73
L47549.1 - 5 - 6 195 73
L47550.1 - 5 - 6 180 73
L47551.1 - 4 - 5 181 73
L47552.1 - 4 - 5 190 73
L47553.1 - 4 - 5 197 73
L47599.1 - 5 - 6 201 73
L47600.1 - 4 - 5 198 73

View Full Product Details